Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Disruptor Beads 0.1 mm 8 oz.

Catalog No. 01804412 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Beads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01-804-412 Beads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-804-412 Supplier OHAUS Supplier No. 83041302
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glass beads of 0.1 mm ideal for disruption of bacteria and DNA.

  • Glass beads of 0.1 mm are designed for use with the Disruptor Genie or TurboMix™ attachment (used with Vortex-Genie™ 2 or Vortex-Genie™ 2T) for optimum disruption of bacteria, yeast, fungi and other cells.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Beads
Model Disruptor Genie™ or TurboMix™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.