Learn More
Chemglass Life Sciences Disruptor Beads, 0.1mm, 8oz/Bottle
SDP
Catalog No. 5000083193
Encompass
0. 1mm for bacteria and 0. 5mm for yeast.
Catalog No. 50-000-83193 Supplier Chemglass Life Sciences Supplier No. CLS1835BG1
0. 1mm for bacteria and 0. 5mm for yeast.