Learn More
Chemglass Life Sciences Accessory: Disruptor Beads, 0.5mL, 8oz Bottle
SDP
Catalog No. 501214910
Encompass
0. 1mm for bacteria and 0. 5mm for yeast.
Catalog No. 50-121-4910 Supplier Chemglass Life Sciences Supplier No. CLS1835BG5
0. 1mm for bacteria and 0. 5mm for yeast.