Learn More
Research Products International Corp Disruptor Beads, Glass, 0.1mm, 375 Grams
SDP
Catalog No. 501367648
Encompass
Spherical lead free soda lime glass beads are available for optimal disruption of cells. Size for use with bacteria - 0.1 mm diameter beads.
Catalog No. 50-136-7648 Supplier Research Products International Corp Supplier No. 155561
Spherical lead free soda lime glass beads are available for optimal disruption of cells. Size for use with bacteria - 0.1 mm diameter beads.