Learn More
Research Products International Corp Disruptor Beads, Glass, 0.5mm, 375 Grams
SDP
Catalog No. 501367649
Encompass
Spherical lead free soda lime glass beads are available for optimal disruption of cells. 0.5 mm diameter beads for use with Yeast/Fungi.
Catalog No. 50-136-7649 Supplier Research Products International Corp Supplier No. 155564
Spherical lead free soda lime glass beads are available for optimal disruption of cells. 0.5 mm diameter beads for use with Yeast/Fungi.