Learn More
Electron Microscopy Sciences Genie™ Media Disruptor with 2.0 ml Microtube Holder, 120 V
SDP
Catalog No. 501899892
Encompass_Preferred
Rapid hands-free disruption of up to 12 microtubes (1.5 ml or 2.0 ml) at one time.
Unique agitation, vortexing, and collision actions for cell disruption and sample resuspension.
Maintain operating speed for up to 15 minutes of continuous operation, even in cold rooms.
Supplied pop-up cup easily attaches for use as a standard single-tube vortex mixer.
Ideal for glass bead procedures using Disruptor Beads. Less costly than ultrasonic units or homogenizes.
Disruptor Bead are spherecal lead-free soda lime glass beads used for disruption of yeast, bacteria, and other samples in combination with Disruptor Genie or TurboMix attachments for Vortex-Genie 2, Vortec-Genie 2T or Digital Vortex-Genie 2. Supplied with 2.0 ml Microtube Holder.
Catalog No. 50-189-9892 Supplier Electron Microscopy Sciences Supplier No. 7240712
Rapid hands-free disruption of up to 12 microtubes (1.5 ml or 2.0 ml) at one time.
Unique agitation, vortexing, and collision actions for cell disruption and sample resuspension.
Maintain operating speed for up to 15 minutes of continuous operation, even in cold rooms.
Supplied pop-up cup easily attaches for use as a standard single-tube vortex mixer.
Ideal for glass bead procedures using Disruptor Beads. Less costly than ultrasonic units or homogenizes.
Disruptor Bead are spherecal lead-free soda lime glass beads used for disruption of yeast, bacteria, and other samples in combination with Disruptor Genie or TurboMix attachments for Vortex-Genie 2, Vortec-Genie 2T or Digital Vortex-Genie 2. Supplied with 2.0 ml Microtube Holder.