Learn More
Chemglass Life Sciences Disruptor Genie, Digital, Universal, 120V
SDP
Catalog No. 5000083094
Encompass
The patented Disruptor Genie provides multi-directional action, which simultaneously agitates and vortexes at high speed, dramatically increasing cell disruption or sample re-suspension efficiency. Maintains operating speed for up to 15 minutes, or continuous operation, even in cold rooms. An affordable alternative to expensive ultrasonic disruptors, it can hold up to (12) 1. 5mL or (12) 2. 0mL micro tubes. In addition, a supplied pop-off cup easily attaches for use as a standard single-tube vortex mixer. Ideal for glass bead procedures using Disruptor Beads. Disrupter beads sold separately: 0.1mL for bacteria and 0.5mL for yeast.
Catalog No. 50-000-83094 Supplier Chemglass Life Sciences Supplier No. CLS1835D25
The patented Disruptor Genie provides multi-directional action, which simultaneously agitates and vortexes at high speed, dramatically increasing cell disruption or sample re-suspension efficiency. Maintains operating speed for up to 15 minutes, or continuous operation, even in cold rooms. An affordable alternative to expensive ultrasonic disruptors, it can hold up to (12) 1. 5mL or (12) 2. 0mL micro tubes. In addition, a supplied pop-off cup easily attaches for use as a standard single-tube vortex mixer. Ideal for glass bead procedures using Disruptor Beads. Disrupter beads sold separately: 0.1mL for bacteria and 0.5mL for yeast.