Learn More
Med Vet International Diss Male, J0539DM, 0.25in NPT, Each
Catalog No. 5000585238
Encompass_Preferred
DISS male x 1/4" male NPT
Catalog No. 50-005-85238 Supplier Med Vet International Supplier No. JOR539DM
DISS male x 1/4" male NPT