Learn More
StatLab™ Dissect Aid™
GSA_VASDP
Catalog No. STL90011
Change view
Click to view available options
Quantity:
2.5 gal.
5 gal.
4 x 1 gal.
Packaging Type:
Bottle
Cube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
STL90011 4 x 1 gal. Bottle
STL900125 2.5 gal. Cube
STL90015 5 gal. Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STL90011 Supplier StatLab™ Supplier No. 90011
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
A fixative which helps make lymph nodes more identifiable during pathology examination.
  • Turns lymph nodes pale to white so they stand out from other tissue, while firming the nodes for touch identification when performing gross examination
  • Dramatically improves lymph node recovery or harvesting
  • Ideal fixative for fatty tissues
  • Formulation has helped in diagnosis for the past 30 years
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Packaging Type Bottle
Quantity 4 x 1 gal.
Formulation Ethyl Alcohol, Acetic Acid, Methyl Alcohol, Formaldehyde, Isopropyl Alcohol