Learn More
StatLab™ Dissect Aid™
Description
- Turns lymph nodes pale to white so they stand out from other tissue, while firming the nodes for touch identification when performing gross examination
- Dramatically improves lymph node recovery or harvesting
- Ideal fixative for fatty tissues
- Formulation has helped in diagnosis for the past 30 years
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Packaging Type | Cube |
| Quantity | 5 gal. |
| Formulation | Ethyl Alcohol, Acetic Acid, Methyl Alcohol, Formaldehyde, Isopropyl Alcohol |