Learn More
Electron Microscopy Sciences DissecTable™ Dissecting Board Base
SDP
Catalog No. 5032244
Encompass_Preferred
Heavy duty base is used to elevate the DissecTable Board to the right height. Polystyrene and stackable. Will not move sideways. Retains excess fluid.
Dimensions: 481 mm x 656 mm x 91 mm.
Catalog No. 50-322-44 Supplier Electron Microscopy Sciences Supplier No. 63309DT
Heavy duty base is used to elevate the DissecTable Board to the right height. Polystyrene and stackable. Will not move sideways. Retains excess fluid.
Dimensions: 481 mm x 656 mm x 91 mm.