Learn More
Electron Microscopy Sciences DissecTable™ Dissecting Board
SDP
Catalog No. 5032243
Encompass_Preferred
Polyethylene, one side has a dimensional scales and six dimensional circles. Other side has two cutting boards half the size with the same dimensional features. Corners have rubber feet.
Dimensions: 575mm x 400mm x 12.5mm
Catalog No. 50-322-43 Supplier Electron Microscopy Sciences Supplier No. 63309DB
Polyethylene, one side has a dimensional scales and six dimensional circles. Other side has two cutting boards half the size with the same dimensional features. Corners have rubber feet.
Dimensions: 575mm x 400mm x 12.5mm