Learn More
StatLab DissecTable Junior
SDP
Catalog No. 22021388
Change view
Click to view available options
Material:
Polyethylene
Catalog No. Material
22-021-388 Polyethylene
1 options
Catalog No. 22-021-388 Supplier StatLab Supplier No. LWC07
Add to Cart
Add to Cart
DissecTable Junior
  • Used to provide a dedicated clean surface for carefully cutting and examining tissue samples under a microscope
Specifications
Product Type Dissection Board
Autoclavable Not Autoclavable
Color White
Material Polyethylene
Disposable Reusable
Sterility Non-sterile