Learn More
Research Products International Corp Dissecting Blade, Stainless Steel, Size 11, Sterile, Individually Wrapped, 100 per Case
SDP
Supplier:  Research Products International Corp 248171
Encompass
Stainless steel blades are made from surgical grade British stainless steel for maximum strength and durability. Blades are sterile and individually wrapped in easy to use peel open pouches.
Catalog No. 50-476-540