Learn More
Research Products International Corp Dissecting Blade, Size 12, 100/cs
SDP
Catalog No. 501977927
Encompass
Stainless steel blades are made from surgical grade British stainless steel for maximum strength and durability. Blades are sterile and individually wrapped in easy to use peel open pouches.
Catalog No. 50-197-7927 Supplier Research Products International Corp Supplier No. 248172
Stainless steel blades are made from surgical grade British stainless steel for maximum strength and durability. Blades are sterile and individually wrapped in easy to use peel open pouches.