Learn More
Electron Microscopy Sciences Dissecting Dish with Plastic Dish
SDP
Catalog No. 50930381
Encompass_Preferred
A silicone pad placed inside a 150 mm x 20 mm deep plastic petri dish. The mat can be removed, washed, and returned to the dish for reuse. The pins come in 10 different coded colors.
Catalog No. 50-930-381 Supplier Electron Microscopy Sciences Supplier No. 70540
A silicone pad placed inside a 150 mm x 20 mm deep plastic petri dish. The mat can be removed, washed, and returned to the dish for reuse. The pins come in 10 different coded colors.