Learn More
Andwin Scientific DISSECTING FORCEPTS 5/PK
SDP
Catalog No. NC9477566
Encompass_Preferred
These dissecting forceps featurea curved design with smooth points. This product is made of stainless steel and has an overall length of 150 millimeters or 6 inches. It is sold in a pack of 5
Catalog No. NC9477566 Supplier Andwin Scientific Supplier No. 25718008
These dissecting forceps featurea curved design with smooth points. This product is made of stainless steel and has an overall length of 150 millimeters or 6 inches. It is sold in a pack of 5