Learn More
Electron Microscopy Sciences Vinyl Mat Only, 13 1/8” x 9 3/8”
SDP
Catalog No. 501929417
Encompass_Preferred
Removable reusable vinyl pad accepts dissecting as well as push-pins to hold down your specimen during dissecting. The pad measures 13 1/8" x 9 3/8" x 2 1/4" thick.
Dimensions: 13 1/8” x 9 3/8”
Thickness: 2-1/4
Catalog No. 50-192-9417 Supplier Electron Microscopy Sciences Supplier No. 63313P
Removable reusable vinyl pad accepts dissecting as well as push-pins to hold down your specimen during dissecting. The pad measures 13 1/8" x 9 3/8" x 2 1/4" thick.
Dimensions: 13 1/8” x 9 3/8”
Thickness: 2-1/4