Learn More
Fine Science Tools Metzenbaum Scissors, Stainless Steel, 14.5 cm
SDP
Supplier:  Fine Science Tools 1401614
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 31mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No
Catalog No. NC9468364