Learn More
Sklar Instruments Econo Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. PK/12.
SDP
Catalog No. 501437595 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Econo Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth, blunt/blunt tips and a length of 5-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7595 Supplier Sklar Instruments Supplier No. 21308
Econo Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Econo Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth, blunt/blunt tips and a length of 5-1/2 inches. They are sold in boxes of 12.