Learn More
Sklar Instruments Sklarhone Metzenbaum Dissecting Scissors, Sklar, 7 in., Blunt/Blunt, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Finger Rings / Dual Black, Stainless Steel. EA/1.
Supplier: Sklar Instruments 153410
Sklarhone Metzenbaum Dissecting Scissors, Sklar, 7 in., Blunt/Blunt, Curved, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Finger Rings / Dual Black, Stainless Steel. Supercut Sklarhone Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures or drains. Supercut Sklarhone scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone instruments are identified by dual black finger rings. This product is curved, has one razor edge, one micro-serrated edge, blunt tips, and an overall length of 7 inches.