Learn More
Cole-Parmer Cole-Parmer Dissecting Scissors, Standard Grade, Sharp Point, Straight, 5.5".
Catalog No. NC0751449 Shop All Cole-Parmer Products
Encompass_Preferred
Standard-grade instruments are manufactured from stainless steel; hardened, tempered, and polished for a high-priced finish without the added cost. These instruments are well-suited for less critical applications.
Autoclavable Note: Not intended for medical applications or patient use.
Catalog No. NC0751449 Supplier Cole-Parmer Supplier No. 1081800
Standard-grade instruments are manufactured from stainless steel; hardened, tempered, and polished for a high-priced finish without the added cost. These instruments are well-suited for less critical applications.
Autoclavable Note: Not intended for medical applications or patient use.