Learn More
Sklar Instruments Econo Metzenbaum-Lahey Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. PK/12.
SDP
Catalog No. 501437607 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Metzenbaum-Lahey Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Econo Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product has straight, smooth blades, blunt tips, and an overall length of 5-1/2 inches. It is sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7607 Supplier Sklar Instruments Supplier No. 21335
Econo Metzenbaum-Lahey Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Econo Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product has straight, smooth blades, blunt tips, and an overall length of 5-1/2 inches. It is sold in boxes of 12.