Learn More
Med Vet International 15-pc. Dissecting Kit, with Case, Each
Catalog No. 502223964
Encompass_Preferred
This comprehensive dissecting set includes:Dissecting Needle, Straight Dissecting Needle, Curved 4 1/2in. Iris Micro Surgical Scissors 5 1/2in. Operating Scissors S/C Halsted Mosquito Forceps 1 Adson Forceps 1 Surgical Probe 1 Micro Graff Forceps, Curved 1 Micro Graff Forceps, Straight #3 Scalpel Handle #4 Scalpel Handle Two #10 Scalpel Blades Two #22 Scalpel Blades
Catalog No. 50-222-3964 Supplier Med Vet International Supplier No. DK15
This comprehensive dissecting set includes:Dissecting Needle, Straight Dissecting Needle, Curved 4 1/2in. Iris Micro Surgical Scissors 5 1/2in. Operating Scissors S/C Halsted Mosquito Forceps 1 Adson Forceps 1 Surgical Probe 1 Micro Graff Forceps, Curved 1 Micro Graff Forceps, Straight #3 Scalpel Handle #4 Scalpel Handle Two #10 Scalpel Blades Two #22 Scalpel Blades