Learn More
Electron Microscopy Sciences EMS SuperCut Dissection Scissors with Curved Blades, 5-1/2" OAL
SDP
Catalog No. 501931409
Encompass_Preferred
These dissection scisssors are made from stainless steel with sharp/blunt tips. Blades have a curve to attain the hard to reach delicate tissue or angle dissections. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings
Blade: Curved
OAL: 5-1/2"
Catalog No. 50-193-1409 Supplier Electron Microscopy Sciences Supplier No. 7299602
These dissection scisssors are made from stainless steel with sharp/blunt tips. Blades have a curve to attain the hard to reach delicate tissue or angle dissections. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings
Blade: Curved
OAL: 5-1/2"