Learn More
Electron Microscopy Sciences EMS Tungsten Carbide Dissection Scissors with Curved Blades Sharp/Sharp, 5-1/2"
SDP
Catalog No. 501931428
Encompass_Preferred
These dissecting scissors have a tungsten carbide cutting edge that provides effortless, yet accurate, cuts through the toughest of surgical or autopsy tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex dissection cases.
Blade: Curved Sharp/Sharp
OAL: 5-1/2"
Catalog No. 50-193-1428 Supplier Electron Microscopy Sciences Supplier No. 7299620
These dissecting scissors have a tungsten carbide cutting edge that provides effortless, yet accurate, cuts through the toughest of surgical or autopsy tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex dissection cases.
Blade: Curved Sharp/Sharp
OAL: 5-1/2"