Learn More
Fine Science Tools Dissector Scissors - Slim Blades
SDP
Catalog No. NC9462540
Encompass
Tip Shape: Angled to Side/ Tips: Sharp-Sharp/ Cutting Edge: 8mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No. Slim blades for cutting tissue.
Catalog No. NC9462540 Supplier Fine Science Tools Supplier No. 1408109
Tip Shape: Angled to Side/ Tips: Sharp-Sharp/ Cutting Edge: 8mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No. Slim blades for cutting tissue.