Learn More
Quality Lab Accessories Ever-Clear-48 Water Preservative, Prevents Bacteria Growth, 48 Single-Dose Containers
Catalog No. NC1843563
Encompass
The EVER-CLEAR-48 Dissolution Bath Water Treatment is a non-hazardous water preservative that prevents bacteria and algae growth while ensuring crystal clear water. With its FDA and EPA approved formula, it simplifies maintenance for dissolution baths by incorporating special cleaning agents and protecting 6-8 vessels for up to 8 weeks. Its environmentally friendly design makes it a reliable and efficient choice for maintaining optimal water quality in laboratory settings.
Catalog No. NC1843563 Supplier Quality Lab Accessories Supplier No. EVERCLEAR48
The EVER-CLEAR-48 Dissolution Bath Water Treatment is a non-hazardous water preservative that prevents bacteria and algae growth while ensuring crystal clear water. With its FDA and EPA approved formula, it simplifies maintenance for dissolution baths by incorporating special cleaning agents and protecting 6-8 vessels for up to 8 weeks. Its environmentally friendly design makes it a reliable and efficient choice for maintaining optimal water quality in laboratory settings.