Learn More
BMEM BIO Dissociation
Supplier:  BMEM BIO 1002
Encompass
BMEM Dissociation is a ready-to-use, chemically defined, animal component-free, and enzyme-free solution specifically optimized for passaging human induced pluripotent stem cells (hiPSCs) in adherent culture.•Gentle & Effective: Formulated with optimized dissociation agents in a BMEM-based buffer to ensure maximal cell survival and viability during passaging.•Workflow Compatible: Extensively validated for use with modern weekend-free and no-change feeding protocols.•System Synergy: Recommended for use with BMEM Human Primed Pluripotent media.•Broad Matrix Compatibility: Validated with leading matrices including Corning Matrigel, Gibco™ Geltrex™, Cultrex™ UltiMatrix, Sigma-Aldrich Laminin, and Recombinant Vitronectin.•Quality Assured: Each lot is performance tested on hiPSCs to ensure consistent biological activity.
Catalog No. NC3932563