Learn More
Bertin Corp Precellys Multi-Tissue Dissociation Kit
SDP
Catalog No. NC3254450
Encompass_Preferred
The Precellys Multi-Tissue Dissociation Kit developed by Bertin Technologies is an innovative and patented solution designed for the obtention of viable single cells from biological samples for research purposes. It enables the obtention of high-quality single cells from complex tissues such as mouse spleen, liver, lung, heart, and thymus and opens the door to downstream single-cell analysis, such as flow cytometry or NGS.
Catalog No. NC3254450 Supplier Bertin Corp Supplier No. D05717.20EA
The Precellys Multi-Tissue Dissociation Kit developed by Bertin Technologies is an innovative and patented solution designed for the obtention of viable single cells from biological samples for research purposes. It enables the obtention of high-quality single cells from complex tissues such as mouse spleen, liver, lung, heart, and thymus and opens the door to downstream single-cell analysis, such as flow cytometry or NGS.