Learn More
Bertin Corp Precellys Dissociation tubes
SDP
Catalog No. NC3254452
Encompass_Preferred
The Precellys Dissociation tubes are 15 mL tubes, sterile, that were developed to dissociate Mouse Heart, Lung, Liver, Spleen, and Thymus tissue.
Catalog No. NC3254452 Supplier Bertin Corp Supplier No. D30717.20EA
The Precellys Dissociation tubes are 15 mL tubes, sterile, that were developed to dissociate Mouse Heart, Lung, Liver, Spleen, and Thymus tissue.