Learn More
Hach Company Dissolved Oxygen AccuVac Ampules, High Range, pk/25
Catalog No. 501996163 Shop All Hach Company Products
Encompass
For determination of Dissolved Oxygen by Hach methods 8166 and 8333. Ranges: 0.3 - 15.0 or 1 - 40 mg/L O₂. Pack of 25 ampules.
Catalog No. 50-199-6163 Supplier Hach Company Supplier No. 2515025
For determination of Dissolved Oxygen by Hach methods 8166 and 8333. Ranges: 0.3 - 15.0 or 1 - 40 mg/L O₂. Pack of 25 ampules.