Learn More
Hach Company Dissolved Oxygen AccuVac Ampules, LR, pk/25
Catalog No. 501996095 Shop All Hach Company Products
Encompass
For determination of low range Dissolved Oxygen by the Indigo Carmine Method, using AccuVac Ampules. Hach Method 8316. Range: 8 - 1000 µg/L O2. Package of 25 ampules
Catalog No. 50-199-6095 Supplier Hach Company Supplier No. 2501025
For determination of low range Dissolved Oxygen by the Indigo Carmine Method, using AccuVac Ampules. Hach Method 8316. Range: 8 - 1000 µg/L O2. Package of 25 ampules