Learn More
Hach Company Dissolved Oxygen 1 Reagent Powder Pillows, 60mL sample, pk/100
Catalog No. 501994483 Shop All Hach Company Products
Encompass
For determination of dissolved oxygen by the Winkler titration method. Pack of 100 powder pillows.
Catalog No. 50-199-4483 Supplier Hach Company Supplier No. 98199
For determination of dissolved oxygen by the Winkler titration method. Pack of 100 powder pillows.