Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Dissolved Oxygen/BOD Testab™ Module
SDPSCI_ED

Catalog No. s81623 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
100 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81623 100 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81623 Supplier LaMotte™ Supplier No. 5889
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simple TesTab™ method for measuring dissolved oxygen.

  • TesTabs™ to perform 100 tests for dissolved oxygen and 100 tests for BOD
  • Easy-to-read diagrammed instructions
  • Test factor flash card with helpful background information
  • Includes: color chart/instruction card and test tubes

Specifications

Grade 5 to 12
Age 10 to 17
Includes Flash card, color chart/instruction card, two test tubes with caps, TesTabs for 100 tests each dissolved oxygen and BOD
Quantity 100 Tests/Kit
Class Size 1 Class
Product Type Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.