Learn More
CHEMetrics™ by AquaPhoenix Dissolved Oxygen Comparator, 0 to 100 ppb

Description
- C-7599 Dissolved Oxygen Comparator is a component of K-7599 Dissolved Oxygen CHEMets Visual Test Kit and is designed to be compared against the color formed in R-7540 Dissolved Oxygen CHEMets Refill.
- The comparator's range is applicable to many industrial applications. Color standards are pink in color and cover a range of 0-100 ppb oxygen.
Specifications
Specifications
| Analytes | Dissolved Oxygen |
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | R-7540 DO Refill |
| Range | 0 to 100 ppb |
| Sterility | Non-sterile |
| Type | DO Comparator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.