Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Dissolved Oxygen Comparator, 1 to 12 ppm
SDP

Catalog No. 13299604
Change view
Click to view available options
For Use With (Equipment):
K-7512
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-299-604 K-7512
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-604 Supplier CHEMetrics™ by AquaPhoenix Supplier No. C7512
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dissolved oxygen comparator designed for comparison against color formed during the reaction. Supports visual analysis of dissolved oxygen test results.

  • C-7512 Dissolved Oxygen Comparator is a component of K-7512 Dissolved Oxygen CHEMets Visual Test Kit and is designed to be compared against the color formed in R-7512 Dissolved Oxygen CHEMets Refill.
  • The comparator's range is applicable to many environmental applications. Color standards are blue in color and cover a range of 1-12 mg/L Oxygen.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-7512
Range 1 to 12 ppm
Type DO Comparator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.