Learn More
FLIR Systems Dissolved Oxygen ExStik™
SCI_ED
Catalog No. S68595
Click to view available options
Accuracy (DO):
0.4mg/L
Catalog No. Accuracy (DO)
S68595 0.4mg/L
1 options
Catalog No. S68595 Supplier FLIR Systems Supplier No. DO600
Specifications
Temperature (Metric) Range 0°C to 50°C
Range (DO) 0 to 20mg/L
Resolution (DO) 0.01mg/L
Temperature (English) Range 32°F to 122°F
Accuracy (DO) 0.4mg/L