Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Dissolved Oxygen Test Kit Reagent Refill
SDPSCI_ED

Catalog No. S45088R Shop All LaMotte Company Products
Change view
Click to view available options
No. of Tests:
50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
S45088R 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S45088R Supplier LaMotte™ Supplier No. R5860
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Five reagents; makes 50 tests at 10 ppm

Learn how to conduct oxygen-level tests for plants and animals with the Dissolved Oxygen Test Kit Reagent Refill.

  • Direct-reading titrator test measures levels of oxygen.
  • Indicates whether a body of water can sustain healthy aquatic plants and animals.
  • Low oxygen levels signal contamination from organic wastes.
  • Measures 0 to 10 ppm in 0.2 ppm increments.
  • Azide modification of Winkler method.
  • Kit Includes: 30 mL Manganous Sulfate Solution, 30 mL Alkaline Potassium Iodide Azide, 30 mL Sulfuric Acid, 60 mL 0.025 N Sodium Thiosulfate, 30 mL Starch Indicator Solution, Direct Reading Titrator, 5-10-12.9-15-20-25 mL Glass Test Tube with Cap, 60 mL Water Sampling Glass Bottle

Specifications

For Use With (Application) Water Testing
For Use With (Equipment) Dissolved Oxygen Kit
Includes 5 Reagents
No. of Tests 50
Type Dissolved Oxygen Kit Reagent Refill
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.