Learn More
LaMotte™ Dissolved Oxygen Sampler
SDPSCI_ED
Catalog No. S94969 Shop All LaMotte Company Products
Change view
Click to view available options
Dissolved oxygen sampler
Catalog No.
S94969 Dissolved oxygen sampler
1 options
Catalog No. S94969 Supplier LaMotte™ Supplier No. 1054DO
Includes
Water sampling body, 60mL sample collection bottle and sounding weight
Simplified dissolved oxygen water sampler designed for use in the field.
  • Removable inner sample bottle will overflow five times to ensure a representative sample
  • Samples can be taken at a controlled depth using a calibrated line
  • Two-pound weight attached to the bottom of the sampling device ensures rapid descent and minimizes drift due to currents
Specifications
For Use With (Application) Environmental education
Includes Water sampling body, 60mL sample collection bottle, sounding weight
Type Dissolved Oxygen Sampler