Learn More
Grainger Dissolved Oxygen Sensor Epoxy/Noryl
Catalog No. 502767674
Encompass_Preferred
The SENSOREX DO6441-T is a dissolved oxygen sensor that measures dissolved oxygen in water. It has a measurement range of 0-20 mg/L (ppm) O2 with an accuracy of ±1% of reading. The sensor uses optical fluorescence quenching technology with a rugged titanium body and IP68 waterproof rating. It has an operating temperature range of 0-50°C, response time of <60 seconds, and expected lifetime of >5 years. The sensor has a standard 4-20 mA output and is designed for online monitoring and control applications in municipal water, wastewater, aquaculture, food & beverage, power & semiconductor industries. It requires no membranes or electrolyte replenishment.
Catalog No. 50-276-7674 Supplier Grainger Supplier No. 16Z043
The SENSOREX DO6441-T is a dissolved oxygen sensor that measures dissolved oxygen in water. It has a measurement range of 0-20 mg/L (ppm) O2 with an accuracy of ±1% of reading. The sensor uses optical fluorescence quenching technology with a rugged titanium body and IP68 waterproof rating. It has an operating temperature range of 0-50°C, response time of <60 seconds, and expected lifetime of >5 years. The sensor has a standard 4-20 mA output and is designed for online monitoring and control applications in municipal water, wastewater, aquaculture, food & beverage, power & semiconductor industries. It requires no membranes or electrolyte replenishment.