Learn More
CHEMetrics™ by AquaPhoenix Dissolved Oxygen Water Quality Visual Test Kit, 5 to 180 ppb
Description
- Kit K-7518 comes in a plastic case and contains everything needed to perform 30 tests: Refill, Comparator, permanent mounting clamp, sampling tube, and instructions.
Specifications
Specifications
| Analytes | Dissolved Oxygen |
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| No. of Tests | 30 |
| Range | 5 to 180 ppb |
| Type | Water Quality Visual Test Kit |