Learn More
Chemglass Life Sciences Adapter, Distillation, 75°, 14/20 Inner Joints, 10/18 Thermometer Joint For 25mm Immersion
SDP
Catalog No. CG102414
Encompass_Preferred
Adapter, Distillation, 75¿, 14/20 Inner Joints, 10/18 Thermometer Joint For 25mm Immersion. Adapter has a standard taper outer joint at the top of the main tube for use with jointed thermometers. The 10/18 size is used with 10/18, 25mm immersion thermometers. With inner joints on the bottom and sidearm at a 75¿ angle to one another.
Catalog No. CG102414 Supplier Chemglass Life Sciences Supplier No. CG102414
Adapter, Distillation, 75¿, 14/20 Inner Joints, 10/18 Thermometer Joint For 25mm Immersion. Adapter has a standard taper outer joint at the top of the main tube for use with jointed thermometers. The 10/18 size is used with 10/18, 25mm immersion thermometers. With inner joints on the bottom and sidearm at a 75¿ angle to one another.