Learn More
Ace Glass, Inc. Congealing temperature distillation apparatus
SDP
Catalog No. NC9021093
Encompass
ACE congealing temperature distillation apparatus, which is similar to that described in U.S. Pharmacopeia, 1995, page 1780. The ACE apparatus is supplied with Ace-Threds in place of rubber stoppers, where practical. A #50 thread at center holds a test tube with a #25 bushing, and the open-top test tube utilizes a rubber stopper to secure a thermometer and wire stirrer. The apparatus has a #7 Ace-Thred thermometer port on one side, and a 22mm I.D. opening on the other side to access the 125mm O.D. x 150mm high cylinder. Applicable to substances that melt down between -20 to 150 degrees C.
Catalog No. NC9021093 Supplier Ace Glass, Inc. Supplier No. 654745
ACE congealing temperature distillation apparatus, which is similar to that described in U.S. Pharmacopeia, 1995, page 1780. The ACE apparatus is supplied with Ace-Threds in place of rubber stoppers, where practical. A #50 thread at center holds a test tube with a #25 bushing, and the open-top test tube utilizes a rubber stopper to secure a thermometer and wire stirrer. The apparatus has a #7 Ace-Thred thermometer port on one side, and a 22mm I.D. opening on the other side to access the 125mm O.D. x 150mm high cylinder. Applicable to substances that melt down between -20 to 150 degrees C.