Learn More
Wilmad Labglass DIST APP MONIER-WILLIAMS SULF
Catalog No. 50804912
Encompass
Distillation Apparatus, Sulfites, Monier-Williams, Complete. Apparatus is used For the determination of sulfites in food and related products. Consists of 3-neck 1000mL boiling flask
Catalog No. 50-804-912 Supplier Wilmad Labglass Supplier No. LG6675100
Distillation Apparatus, Sulfites, Monier-Williams, Complete. Apparatus is used For the determination of sulfites in food and related products. Consists of 3-neck 1000mL boiling flask