Learn More
Wilmad Labglass DIST APP SHORT PATH COMP
Supplier:  Wilmad Labglass LG6315098
Encompass_Preferred
Distilling Apparatus, Short Path, Complete. Apparatus consists of 250mL 2-neck Flask, short path Distilling Head, distributing adapter, three 50mL receiving flasks, 75mm immersion thermometer, bleed-in adapter
Catalog No. 50-805-877