Learn More
Wilmad Labglass DIST APP VOLATILE OIL HTW COMP
Catalog No. 50805162
Encompass
Distilling Apparatus, Volatile Oils, For Oils Heavier Than Water, Complete. Apparatus consists of a 250mL long neck Flask, cold finger Condenser, Clevenger-type trap w/ a [ST] 24/40 Outer joint at top and an inner joint of same size
Catalog No. 50-805-162 Supplier Wilmad Labglass Supplier No. LG6656100
Add to Cart
Add to Cart
Distilling Apparatus, Volatile Oils, For Oils Heavier Than Water, Complete. Apparatus consists of a 250mL long neck Flask, cold finger Condenser, Clevenger-type trap w/ a [ST] 24/40 Outer joint at top and an inner joint of same size