Learn More
Wilmad Labglass Distilling Apparatus, for Oils Lighter than Water
Catalog No. 50805161
Encompass
Distilling Apparatus, Volatile Oils, For Oils Lighter Than Water, Complete. Apparatus consists of a long neck Flask, cold finger Condenser, Clevenger-type trap w/ a [ST] 24/40 Outer joint at top and an inner Joint of same size at bottom
Catalog No. 50-805-161 Supplier Wilmad Labglass Supplier No. LG6655100
Distilling Apparatus, Volatile Oils, For Oils Lighter Than Water, Complete. Apparatus consists of a long neck Flask, cold finger Condenser, Clevenger-type trap w/ a [ST] 24/40 Outer joint at top and an inner Joint of same size at bottom