Learn More
Wilmad Labglass DIST COL HEMPEL 25X300 29/42
Catalog No. 50801495
Encompass
Distilling Column, Hemple, Jacketed, Silvered. Column ID: 25mm; Length For Packing Material: 300mm; Joint Size: 29/42. Similar to LG-5791-series, but w/ jacket completely silvered except For two narrow clear strips 180degree apart.
Catalog No. 50-801-495 Supplier Wilmad Labglass Supplier No. LG5792140
Distilling Column, Hemple, Jacketed, Silvered. Column ID: 25mm; Length For Packing Material: 300mm; Joint Size: 29/42. Similar to LG-5791-series, but w/ jacket completely silvered except For two narrow clear strips 180degree apart.