Learn More
Wilmad Labglass DIST COL VIGREUX 200MM 24/40
Supplier:  Wilmad Labglass LG5890102
Encompass_Preferred
Distilling Column, Vigreux. Effective Length: 200mm; Joint Size: [ST] 24/40. Column indentations, alternating horizontal and downward, improve refluxing and promote separation of volatile mixtures.
Catalog No. 50-807-034